missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoimidazole, 96%, Thermo Scientific Chemicals

Product Code. 15448357
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448357 1 g 1g
15458357 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448357 Supplier Thermo Scientific Alfa Aesar Supplier No. H51868.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3034-62-6
Molecular Formula C3H3IN2
Molecular Weight (g/mol) 193.975
MDL Number MFCD00159701
InChI Key MHHDMDLNVVCTAJ-UHFFFAOYSA-N
Synonym 2-iodoimidazole, iodoimidazole, 1h-imidazole, 2-iodo, sftheavililuh@, 2-iodoimidazole hcl, pubchem9423, acmc-1aies, 2-iodo imidazole
PubChem CID 2773468
IUPAC Name 2-iodo-1H-imidazole
SMILES C1=CN=C(N1)I

Specifications

Melting Point 190°C to 191°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 193.96
Percent Purity 96%
Chemical Name or Material 2-Iodoimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.