missing translation for 'onlineSavingsMsg'
Learn More

2-Iodopropane, 98+%, stab. with copper

Product Code. 11339599
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. Quantity unitSize
11339599 25 g 25g
11477347 100 g 100g
11487347 500 g 500g
3 options
This item is not returnable. View return policy

Product Code. 11339599

Brand: Thermo Scientific Alfa Aesar A15066.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-30-9 | C3H7I | 169.993 g/mol

2-Iodopropane is used to prepare butyric and isobutyric acid, as a solvent, also used in medicine and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodopropane is used to prepare butyric and isobutyric acid, as a solvent, also used in medicine and organic synthesis.

Solubility
Soluble in water(1.4g/L).

Notes
Light Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-30-9
Molecular Formula C3H7I
Molecular Weight (g/mol) 169.993
MDL Number MFCD00001071
InChI Key FMKOJHQHASLBPH-UHFFFAOYSA-N
Synonym isopropyl iodide, propane, 2-iodo, 2-propyl iodide, sec-propyl iodide, isopropyliodide, 2-jodpropan, 2-jodpropan czech, 2-iodo-propane, i-propyl iodide, unii-67k05opz0e
PubChem CID 6362
IUPAC Name 2-iodopropane
SMILES CC(C)I

Specifications

Melting Point -90°C
Density 1.701
Boiling Point 88°C to 90°C
Flash Point 42°C (107°F)
Assay Percent Range 98%, stabilized with copper
Odor Recognizable
Refractive Index 1.4982
Quantity 25 g
UN Number UN2392
Beilstein 1098244
Sensitivity Light sensitive
Merck Index 14,5214
Solubility Information Soluble in water(1.4g/L).
Formula Weight 169.99
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 2-Iodopropane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.