missing translation for 'onlineSavingsMsg'
Learn More

2-Iodopropane, 98+%, stab. with copper

Product Code. 11477347
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477347 100 g 100g
11339599 25 g 25g
11487347 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11477347 Supplier Thermo Scientific Alfa Aesar Supplier No. A15066.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 75-30-9 | C3H7I | 169.993 g/mol

2-Iodopropane is used to prepare butyric and isobutyric acid, as a solvent, also used in medicine and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodopropane is used to prepare butyric and isobutyric acid, as a solvent, also used in medicine and organic synthesis.

Solubility
Soluble in water(1.4g/L).

Notes
Light Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-30-9
Molecular Formula C3H7I
Molecular Weight (g/mol) 169.993
MDL Number MFCD00001071
InChI Key FMKOJHQHASLBPH-UHFFFAOYSA-N
Synonym isopropyl iodide, propane, 2-iodo, 2-propyl iodide, sec-propyl iodide, isopropyliodide, 2-jodpropan, 2-jodpropan czech, 2-iodo-propane, i-propyl iodide, unii-67k05opz0e
PubChem CID 6362
IUPAC Name 2-iodopropane
SMILES CC(C)I

Specifications

Melting Point °C to 90°C
Density 1.701
Boiling Point 88°C to 90°C
Flash Point 42°C (107°F)
Assay Percent Range 98%, stabilized with copper
Refractive Index 1.4982
Quantity 100 g
UN Number UN2392
Beilstein 1098244
Sensitivity Light sensitive
Merck Index 14,5214
Solubility Information Soluble in water(1.4g/L).
Formula Weight 169.99
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 2-Iodopropane, stabilized with copper
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Iodopropane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Iodopropane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.