missing translation for 'onlineSavingsMsg'
Learn More

2-Iodopropane, 98+%, stabilized

Product Code. 10011880 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10011880 100 mL Glass bottle 100mL
10124180 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10011880 Supplier Thermo Scientific Acros Supplier No. 164961000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 75-30-9 | C3H7I | 169.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-30-9 , 7440-50-8
Molecular Formula C3H7I
Molecular Weight (g/mol) 169.99
MDL Number MFCD00001071
InChI Key FMKOJHQHASLBPH-UHFFFAOYSA-N
Synonym isopropyl iodide, propane, 2-iodo, 2-propyl iodide, sec-propyl iodide, isopropyliodide, 2-jodpropan, 2-jodpropan czech, 2-iodo-propane, i-propyl iodide, unii-67k05opz0e
PubChem CID 6362
IUPAC Name 2-iodopropane
SMILES CC(C)I

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -90.0°C
Color Colorless to Yellow
Density 1.7000g/mL
Boiling Point 90.0°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHI
Refractive Index 1.4972 to 1.4992
Quantity 100 mL
Beilstein 01, 114
Merck Index 15, 5260
Specific Gravity 1.7
Solubility Information Solubility in water: slightly soluble. Other solubilities: miscible with chloroform,ehter,alcohol and,benzene
Stabilizer copper
Formula Weight 169.99
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2-Iodopropane, 98+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Iodopropane
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.