missing translation for 'onlineSavingsMsg'
Learn More

2-Iodopropane (Stabilized with Copper), TRC

Product Code. 30004284
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004284 10 g 10g
30004146 50 g 50g
30004030 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004284 Supplier Toronto Research Chemicals Supplier No. TRCI71879510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-30-9
Molecular Formula C3 H7 I
Molecular Weight (g/mol) 169.99
Synonym Propane, 2-iodo- (8CI, 9CI, ACI), 2-Iodopropane (ACI), 1-Methylethyl iodide, 2-Propyl iodide, Isopropyl iodide, sec-Propyl iodide
IUPAC Name 2-iodopropane
SMILES CC(C)I

Specifications

Quantity 10 g
InChI Formula InChI=1S/C3H7I/c1-3(2)4/h3H,1-2H3
Formula Weight 169.9592
Chemical Name or Material 2-Iodopropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.