missing translation for 'onlineSavingsMsg'
Learn More

2-Iodopyrazine, 95%

Product Code. 11396699
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11396699 5 g 5g
11386699 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11396699 Supplier Thermo Scientific Alfa Aesar Supplier No. H26244.06
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 32111-21-0 | C4H3IN2 | 205.986 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32111-21-0
Molecular Formula C4H3IN2
Molecular Weight (g/mol) 205.986
MDL Number MFCD01319019
InChI Key OYWPFIUVDKHHGQ-UHFFFAOYSA-N
Synonym iodopyrazine, 2-iodo-pyrazine, 2-iodo-1,3-diazine, iodo-pyrazine, 2-iodo pyrazine, pyrazine, 2-iodo, acmc-1ad2s
PubChem CID 642841
IUPAC Name 2-iodopyrazine
SMILES C1=CN=C(C=N1)I

Specifications

Density 2.086
Boiling Point 109°C to 110°C (34 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.655
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 205.98
Percent Purity 95%
Chemical Name or Material 2-Iodopyrazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.