missing translation for 'onlineSavingsMsg'
Learn More

2-Iodothiophene, 98+%

Product Code. 10226630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
Packaging:
Glass bottle
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10226630 10 mL Glass bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10226630 Supplier Thermo Scientific Acros Supplier No. 187880100
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3437-95-4 | C4H3IS | 210.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3437-95-4
CAS Min % 98.0
CAS Max % 100.0
Melting Point -40.0°C
Color Brown-Red to Green-Yellow
Density 1.9000g/mL
Boiling Point 181.0°C to 185.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Molecular Formula C4H3IS
Refractive Index 1.6510 to 1.6530
MDL Number MFCD00005424
Quantity 10 mL
Beilstein 17, 34
Specific Gravity 1.9
Synonym thiophene, 2-iodo, 2-thienyl iodide, 2-iodo-thiophene, .alpha.-iodothiophene, iodothiophene, thiophene, iodo, alpha-iodothiophene, a-iodothiophene, thiophene,iodo, 2-iodo thiophene
InChI Key ROIMNSWDOJCBFR-UHFFFAOYSA-N
SMILES C1=CSC(=C1)I
IUPAC Name 2-iodothiophene
Molecular Weight (g/mol) 210.03
PubChem CID 18921
Formula Weight 210.03
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2-Iodothiophene, 98+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Iodothiophene
Signal Word
  • Danger||Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.