missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodoxybenzoic acid, stabilized
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
oxidation reagent
Specifications
Specifications
| Melting Point | 250.0°C |
| Color | White |
| Packaging | Glass bottle |
| Quantity | 10 g |
| Merck Index | 15, 5091 |
| Solubility Information | Solubility in water: slightly soluble |
| Formula Weight | 280.01 |
| Percent Purity | 39 to 50% as 2-Iodoxybenzoic acid |
| Physical Form | Powder, Prills or Agglomerates |
| Chemical Name or Material | 2-Iodoxybenzoic acid |
Safety and Handling
Product Identifier
- 2-Iodoxybenzoic acid, stabilized
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity Category 1
- Specific target organ toxicity Category 3
- H318-Causes serious eye damage.
- H372-Causes damage to organs.
- H335-May cause respiratory irritation.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction