missing translation for 'onlineSavingsMsg'
Learn More

2-Isobutyl-3-methoxypyrazine, 99%, Thermo Scientific™

Product Code. 10687011 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10687011 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10687011 Supplier Thermo Scientific Acros Supplier No. 412690010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 24683-00-9
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C9H14N2O
Refractive Index 1.4912 to 1.4932
MDL Number MFCD00006128
Quantity 1 g
Synonym 2-isobutyl-3-methoxypyrazine, 3-isobutyl-2-methoxypyrazine, 2-methoxy-3-2-methylpropyl pyrazine, 2-methoxy-3-isobutylpyrazine, 2-methoxy-3-isobutyl pyrazine, 1dzk, pyrazine, 2-methoxy-3-2-methylpropyl, unii-s327o0t12o, ibmp, 3-methoxy-2-isobutylpyrazine
InChI Key UXFSPRAGHGMRSQ-UHFFFAOYSA-N
SMILES COC1=NC=CN=C1CC(C)C
IUPAC Name 2-methoxy-3-(2-methylpropyl)pyrazine
Molecular Weight (g/mol) 166.22
PubChem CID 32594
Formula Weight 166.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Isobutyl-3-methoxypyrazine, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Isobutyl-3-methoxypyrazine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.