missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 90%

Product Code. 10077899 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10077899 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10077899 Supplier Thermo Scientific Acros Supplier No. 430450050
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 126726-62-3 | C9H17BO2 | 168.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 126726-62-3
CAS Min % 88.0
CAS Max % 100.0
Color Brown to Colorless
Density 0.8800g/mL
Boiling Point 47.0°C (9.0 Mbar)
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Molecular Formula C9H17BO2
MDL Number MFCD08276843
Quantity 5 g
Specific Gravity 0.88
Synonym 4,4,5,5-tetramethyl-2-prop-1-en-2-yl-1,3,2-dioxaborolane, isopropenylboronic acid pinacol ester, 2-isopropenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-isopropenylboronic acid pinacol ester, 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl isopropene, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-1-methylethenyl, 4,4,5,5-tetramethyl-2-1-methylethenyl-1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-isopropenyl-1,3,2-dioxaborolane, isopropenylboronic acid pinacol ester, stabilized with phenothiazine, pubchem17392
InChI Key SVSUYEJKNSMKKW-UHFFFAOYSA-N
SMILES CC(=C)B1OC(C)(C)C(C)(C)O1
IUPAC Name 4,4,5,5-tetramethyl-2-prop-1-en-2-yl-1,3,2-dioxaborolane
Stabilizer 1% phenothiazine
Molecular Weight (g/mol) 168.04
PubChem CID 10997426
Formula Weight 168.04
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 2-Isopropenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Isopropenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.