missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropyl-d7 Thioxanthone, TRC

Product Code. 30394922
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394922 10 mg
30399603 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30394922 Supplier Toronto Research Chemicals Supplier No. TRCI87350210MG
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1173019-24-3
Molecular Formula C16H7D7OS
Molecular Weight (g/mol) 261.39
Synonym 2-(1-Methylethyl-d7)-9H-thioxanthen-9-one, 2-(Isopropyl-d7)-9H-thioxanthen-2-one, 2-(Isopropyl-d7)thioxanthen-9-one, ITX-d7, Photocure ITX-P-d7, Quantacure 597-d7, Quantacure 659-d7, Quantacure ITX-d7, Speedcure ITX-d7
SMILES O=C1C2=C(C=CC(C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=C2)SC3=CC=CC=C31

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H14OS/c1-10(2)11-7-8-15-13(9-11)16(17)12-5-3-4-6-14(12)18-15/h3-10H,1-2H3/i1D3,2D3,10D
Chemical Name or Material 2-Isopropyl-d7 Thioxanthone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.