missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropyl Thioxanthone, TRC

Product Code. 17989354
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17989354 5 g 5g
17958694 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17989354 Supplier Toronto Research Chemicals Supplier No. TRCI8735005G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5495-84-1
Molecular Formula C16 H14 O S
Molecular Weight (g/mol) 254.35
Synonym 2-(1-Methylethyl)-9H-thioxanthen-9-one, Thioxanthen-9-one, 2-isopropyl- (7CI,8CI), 2-(1-Methylethyl)-9H-thioxanthen-9-one, 2-ITX, 2-Isopropyl-9H-thioxanthen-9-one, 2-Isopropylthioxanthen-9-one, 2-Isopropylthioxanthenone, 2-Isopropylthioxanthone, ITX, JRCure ITX, Omnirad ITX, Photocure ITX-P, Quantacure 597, Quantacure 659, Quantacure ITX, Speedcure ITX
IUPAC Name 2-propan-2-ylthioxanthen-9-one
SMILES CC(C)c1ccc2Sc3ccccc3C(=O)c2c1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C16H14OS/c1-10(2)11-7-8-15-13(9-11)16(17)12-5-3-4-6-14(12)18-15/h3-10H,1-2H3
Formula Weight 254.0765
Chemical Name or Material 2-Isopropylthioxantone ITX
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.