missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropylnaphthalene, 96%

Product Code. 11432138
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11432138 25 g 25g
11422138 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11432138 Supplier Thermo Scientific Alfa Aesar Supplier No. A15153.14
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2027-17-0 | C13H14 | 170.255 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2027-17-0
Molecular Formula C13H14
Molecular Weight (g/mol) 170.255
MDL Number MFCD00014322
InChI Key TVYVQNHYIHAJTD-UHFFFAOYSA-N
Synonym 2-isopropylnaphthalene, 2-iso-propylnaphthalene, naphthalene, 2-isopropyl, naphthalene, 2-1-methylethyl, 2-1-methylethyl naphthalene, beta-isopropylnaphthalene, unii-4dtr9un8sr, 4dtr9un8sr, .beta.-isopropylnaphthalene, 2-methylethyl naphthalene
PubChem CID 16238
IUPAC Name 2-propan-2-ylnaphthalene
SMILES CC(C)C1=CC2=CC=CC=C2C=C1

Specifications

Melting Point 14°C
Density 0.975
Boiling Point 268°C
Flash Point 122°C (252°F)
Refractive Index 1.587
Quantity 25 g
Formula Weight 170.26
Percent Purity 96%
Chemical Name or Material 2-Isopropylnaphthalene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.