missing translation for 'onlineSavingsMsg'
Learn More

2-Keto-D-gluconic acid, hemicalcium salt monohydrate, >98%, MP Biomedicals™

Product Code. 11473050 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473050 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11473050 Supplier MP Biomedicals Supplier No. 0210036510
  • ONLINE EXCLUSIVE PRICE
    1250.00 €
    1182.30 € / 10g
    Save 67.70 €
    5% Off
 Restricted Item
Estimated Shipment: 13-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

2-Keto-D-gluconic Acid hemicalcium salt is Hemicalcium Salt of 2-Keto-D-gluconic acid. It is produced from D-glucose

Chemical Identifiers

CAS 3470-37-9
Molecular Formula C12H20CaO14
Molecular Weight (g/mol) 428.356
MDL Number MFCD00149104
InChI Key QACANIDDQFGAJU-JQTJLMCZSA-N
Synonym calcium 2-keto-d-gluconate
PubChem CID 131852723
IUPAC Name calcium;(3S,4R,5R)-3,4,5,6-tetrahydroxy-2-oxohexanoic acid
SMILES C(C(C(C(C(=O)C(=O)O)O)O)O)O.C(C(C(C(C(=O)C(=O)O)O)O)O)O.[Ca]

Specifications

Color White
Assay Percent Range >98%
Quantity 10 g
Formula Weight 213.2
Physical Form Powder
Chemical Name or Material 2-Keto-D-gluconic acid, hemicalcium salt
compliance-icons
Product Identifier
  • 2-Keto-D-gluconic Acid Hemicalcium Salt Monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 2-Keto-D-gluconic Acid Hemicalcium SaltMonohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.