missing translation for 'onlineSavingsMsg'
Learn More

2-Ketoglutaric acid, disodium salt, dihydrate, 99%

Product Code. 10277500 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10277500 50 g Glass bottle 50g
10623373 5 g Glass bottle 5g
10095993 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10277500 Supplier Thermo Scientific Acros Supplier No. 271250500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 305-72-6 | C5H4Na2O5·2H2O | 226.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 305-72-6
Molecular Formula C5H4Na2O5·2H2O
Molecular Weight (g/mol) 226.1
MDL Number MFCD00150702
InChI Key YBGBJYVHJTVUSL-UHFFFAOYSA-L
Synonym disodium 2-oxoglutarate, unii-flp7p4rm46, disodium 2-oxopentanedioate, flp7p4rm46, pentanedioic acid, 2-oxo-, disodium salt, 2-oxoglutaric acid disodium salt, 2-ketoglutaric acid disodium salt, alpha-ketoglutaric acid sodium salt, disodium ? ketoglutarate, disodium oxoglurate
PubChem CID 31040
IUPAC Name disodium;2-oxopentanedioate
SMILES C(CC(=O)[O-])C(=O)C(=O)[O-].[Na+].[Na+]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color White
Loss on Drying 12% to 16%
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula NaOOCCH2CH2C(O)COONa·2H2O
Quantity 50 g
Beilstein 03, 1813
Solubility Information Solubility in water: soluble in water
Formula Weight 226.1
Percent Purity 99%
Physical Form Crystalline Powder or Granules
Chemical Name or Material 2-Ketoglutaric acid, disodium salt, dihydrate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.