missing translation for 'onlineSavingsMsg'
Learn More

2-Linoleoyl-rac-glycerol-d5, TRC

Product Code. 30477963
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477963 1 mg 1mg
30473127 10 mg 10mg
30472912 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30477963 Supplier Toronto Research Chemicals Supplier No. TRCL4679621MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Linoleoyl-rac-glycerol-d5
Molecular Formula C21H33D5O4
Quantity 1 mg
Synonym (9Z,12Z)-9,12-Octadecadienoic Acid 2-Hydroxy-1-(hydroxymethyl)ethyl Ester-d5, 2-Linoleoylglycerol-d5, 2-Monolinolein-d5, 2-Monolinoleoylglycerol-d5, β-Monolinolein-d5
InChI Formula InChI=1S/C21H38O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-20(18-22)19-23/h6-7,9-10,20,22-23H,2-5,8,11-19H2,1H3/b7-6-,10-9-/i18D2,19D2,20D
SMILES O=C(CCCCCCC/C=C\C/C=C\CCCCC)OC(C([2H])([2H])O)([2H])C([2H])([2H])O
Molecular Weight (g/mol) 359.55
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.