missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-1,3,4-thiadiazole, 98%

Product Code. 11465127
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465127 50 g 50g
11455127 10 g 10g
11475127 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11465127 Supplier Thermo Scientific Alfa Aesar Supplier No. B22556.18
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18686-82-3 | C2H2N2S2 | 118.172 g/mol

2-Mercapto-1,3,4-thiadiazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercapto-1,3,4-thiadiazole is used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18686-82-3
Molecular Formula C2H2N2S2
Molecular Weight (g/mol) 118.172
MDL Number MFCD00040404
InChI Key JLAMDELLBBZOOX-UHFFFAOYSA-N
Synonym 2-mercapto-1,3,4-thiadiazole, 1,3,4-thiadiazole-2-thiol, 2-mercapto-1,3,4-thiadiazol, 1,3,4-thiadiazole-2 3h-thione, 1,3,4-thiadiazol-2-yl hydrosulfide, 5-mercapto-1,3,4-thiadiazole, mtd:2-mereapto-1,3,4-thiadiazole, acmc-20ansd, pubchem10119, ksc177e6p
PubChem CID 3034302
IUPAC Name 3H-1,3,4-thiadiazole-2-thione
SMILES C1=NNC(=S)S1

Specifications

Melting Point 142°C to 144°C
Quantity 50 g
Solubility Information Soluble in methanol.
Formula Weight 118.17
Percent Purity 98%
Chemical Name or Material 2-Mercapto-1,3,4-thiadiazole
compliance-icons
Product Identifier
  • 2-Mercapto-1,3,4-thiadiazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1,3,4-Thiadiazole-2(3H)-thione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.