missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-1-methylimidazole, 98%

Product Code. 10041260 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10041260

Brand: Thermo Scientific Acros 125501000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Mercapto-1-methylimidazole, CAS # 60-56-0, also known as methimazole, is a thyroid hormone antagonist used to study the treatment of thyroid disorders. | CAS: 60-56-0 | C4H6N2S | 114.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • 2-Mercapto-1-methylimidazole, also known as methimazole, is an antithyroid medication
  • Methimazole works by inhibiting the enzyme thyroid peroxidase (TPO), which is responsible for synthesizing the thyroid hormones triiodothyronine (T3) and thyroxine (T4) from the precursor thyroglobulin

Applications

  • Methimazole has been used in laboratory settings to model olfactory epithelial degeneration
  • May be used to study disruptions in iodide metabolism
  • Methimazole has been used in Pharmacokinetics studies in the laboratory
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-56-0
Molecular Formula C4H6N2S
Molecular Weight (g/mol) 114.17
MDL Number MFCD00179321
InChI Key PMRYVIKBURPHAH-UHFFFAOYSA-N
Synonym methimazole, thiamazole, 2-mercapto-1-methylimidazole, tapazole, mercazolyl, 1-methylimidazole-2-thiol, mercazole, metazolo, methimazol, thiamazol
PubChem CID 1349907
ChEBI CHEBI:50673
IUPAC Name 3-methyl-1H-imidazole-2-thione
SMILES CN1C=CNC1=S

Specifications

Melting Point 140.0°C to 145.0°C
Color Cream to White
Boiling Point 280.0°C
Flash Point >100°C
Loss on Drying 1% (105°C, 2 hrs) max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 100 g
Beilstein 24,17
Merck Index 15,6043
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and acetone,sparingly soluble in ether,petr. ether,benzene
Formula Weight 114.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercapto-1-methylimidazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Mercapto-1-methylimidazole
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.