missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-5-methoxybenzimidazole, 99%

Product Code. 11458957
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458957 5 g 5g
11468957 25 g 25g
11478957 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458957 Supplier Thermo Scientific Alfa Aesar Supplier No. B20570.06
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 37052-78-1 | C8H8N2OS | 180.23 g/mol

2-Mercapto-5-methoxybenzimidazole is used in the preparation of COX-2 inhibitors. It is also employed in the preparation of compounds such as 5-methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfonyl]-1H-benzimidazole and 5-methoxy-2-[[(4-methoxy-3,5-dimethylpyridin-2-yl-1-oxide)methyl]sulfonyl]-1H-benzimidazole. Further, it acts as a hetercocyclic building block and serves as an intermediate of drug omeprazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercapto-5-methoxybenzimidazole is used in the preparation of COX-2 inhibitors. It is also employed in the preparation of compounds such as 5-methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfonyl]-1H-benzimidazole and 5-methoxy-2-[[(4-methoxy-3,5-dimethylpyridin-2-yl-1-oxide)methyl]sulfonyl]-1H-benzimidazole. Further, it acts as a hetercocyclic building block and serves as an intermediate of drug omeprazole.

Solubility
Soluble in dimethyl sulfoxide. Slightly soluble in methanol. Insoluble in water.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents, acids, bases and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37052-78-1
Molecular Formula C8H8N2OS
Molecular Weight (g/mol) 180.23
MDL Number MFCD00134581
InChI Key KOFBRZWVWJCLGM-UHFFFAOYSA-N
Synonym 5-methoxy-2-mercaptobenzimidazole, 2-mercapto-5-methoxybenzimidazole, 5-methoxy-1h-benzo d imidazole-2-thiol, 5-methoxy-2-benzimidazolethiol, 5-methoxy-1h-benzimidazole-2-thiol, 5-methoxy-1h-1,3-benzodiazole-2-thiol, 2-mercapto-5-methoxy-1h-benzimidazole, 5-methoxy-1h-benzoimidazole-2-thiol, 1,3-dihydro-5-methoxy-2h-benzimidazole-2-thione, 5-methoxybenzimidazole-2-thiol
PubChem CID 665603
SMILES COC1=CC=C2NC(=S)NC2=C1

Specifications

Melting Point 255°C to 259°C
Quantity 5 g
Beilstein 3605123
Solubility Information Soluble in dimethyl sulfoxide. Slightly soluble in methanol. Insoluble in water.
IUPAC Name 5-methoxy-2,3-dihydro-1H-1,3-benzodiazole-2-thione
Formula Weight 180.23
Percent Purity 99%
Chemical Name or Material 2-Mercapto-5-methoxybenzimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.