missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-5-methylbenzimidazole, 98%

Product Code. 11463494
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463494 100 g 100g
11399589 5 g 5g
11453494 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463494 Supplier Thermo Scientific Alfa Aesar Supplier No. A11192.22
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 27231-36-3 | C8H8N2S | 164.23 g/mol

2-Mercapto-5-methylbenzimidazole is used as an additive in liquid paraffin. And also used in electrochemical techniques, corrosion inhibitors for steel under highly acidic conditions and pointed out that the effectiveness of the heterocyclic compound can be intimately associated to its molecular structure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercapto-5-methylbenzimidazole is used as an additive in liquid paraffin. And also used in electrochemical techniques, corrosion inhibitors for steel under highly acidic conditions and pointed out that the effectiveness of the heterocyclic compound can be intimately associated to its molecular structure.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27231-36-3
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.23
MDL Number MFCD00010617
InChI Key CWIYBOJLSWJGKV-UHFFFAOYSA-N
Synonym 2-mercapto-5-methylbenzimidazole, 5-methyl-1h-benzo d imidazole-2 3h-thione, 5-methyl-1h-benzimidazole-2-thiol, 5-methyl-2-benzimidazolethiol, 5-methyl-2-mercaptobenzimidazole, 5-methyl-1h-1,3-benzodiazole-2-thiol, 2h-benzimidazole-2-thione, 1,3-dihydro-5-methyl, 1,3-dihydro-5-methyl-2h-benzimidazole-2-thione, 5-methyl-1h-benzoimidazole-2-thiol, 5-methylbenzimidazole-2-thiol
PubChem CID 712373
IUPAC Name 5-methyl-1,3-dihydrobenzimidazole-2-thione
SMILES CC1=CC=C2NC(=S)NC2=C1

Spécification

Melting Point ∼295°C (decomposition)
Quantity 100 g
Beilstein 5083
Solubility Information Insoluble in water.
Formula Weight 164.23
Percent Purity 98%
Chemical Name or Material 2-Mercapto-5-methylbenzimidazole

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.