missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptobenzimidazole, 98%

Product Code. 10477320 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
1kg
250g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10477320 5 g Glass bottle 5g
10487320 100 g Plastic bottle 100g
10275170 250 g Plastic bottle 250g
10397050 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10477320 Supplier Thermo Scientific Acros Supplier No. 125440050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 583-39-1 | C7H6N2S | 150.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-39-1
Molecular Formula C7H6N2S
Molecular Weight (g/mol) 150.20
MDL Number MFCD00466107
InChI Key YHMYGUUIMTVXNW-UHFFFAOYSA-N
Synonym 2-mercaptobenzimidazole, 1h-benzoimidazole-2-thiol, 2-benzimidazolethiol, 1h-benzimidazole-2-thiol, 1h-benzo d imidazole-2-thiol, o-phenylenethiourea, antioxidant mb, mercaptobenzimidazole, 2-mercaptobenzoimidazole, antiegene mb
PubChem CID 707035
SMILES S=C1NC2=CC=CC=C2N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 302°C to 307°C
Color Beige to White
Boiling Point 451°C
Flash Point >250°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 24, 119
Merck Index 15, 1084
Solubility Information Solubility in water: 2.6g/L (23°C). Other solubilities: soluble in acetone,ethyl acetate and ethanol
IUPAC Name 1,3-dihydrobenzimidazole-2-thione
Formula Weight 150.2
Percent Purity 98%
Insoluble Matter 0.3% max. (in NaOH)
Physical Form Powder
Chemical Name or Material 2-Mercaptobenzimidazole, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Mercaptobenzimidazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.