missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptobenzimidazole, TRC

Product Code. 30138540
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138540 5 g 5g
30003854 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138540 Supplier Toronto Research Chemicals Supplier No. TRCM2251905G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-39-1
Molecular Formula C7 H6 N2 S
Molecular Weight (g/mol) 150.2
Synonym 1,3-Dihydro-2H-benzimidazole-2-thione, 2-Benzimidazolethiol (8CI), 2-Benzimidazolinethione (6CI,7CI), 1,3-Dihydro-2H-benzimidazole-2-thione, 1,3-Dihydro-2H-benzoimidazole-2-thione, 1H-Benzimidazole-2-thiol, 1H-Benzo[d]imidazole-2(3H)-thione, 1H-Benzo[d]imidazole-2-thiol, 2,3-Dihydro-1H-benzimidazole-2-thione, 2-Benzimidazolethione, 2-Mercapto-1H-benzimidazole, 2-Mercaptobenzimidazole, 2-Sulfhydrylbenzimidazole, 2-Thiobenzimidazole, AO-MB, ASM MB, Antage MB, Antigene MB, Antioxidant MB, Antioxidant MB/K, Benzimidazolin-2-thione, Curekind MBI/B, G 580, MB, MB/K, MBI 80, NSC 186246, NSC 21414, Nocrac MB, Nonflex MB, Permanax 21, Rhenogran MBI 80, Sandant MB, Sumilizer MB, Vulkanox MB, Vulkanox MBI, Vulkanox MG, o-Phenylenethiourea, 1H-Benzimidazole-2-thiol, Lansoprazole Imp. E (EP)
IUPAC Name 1H-benzimidazole-2-thiol
SMILES Sc1nc2ccccc2[nH]1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H6N2S/c10-7-8-5-3-1-2-4-6(5)9-7/h1-4H,(H2,8,9,10)
Formula Weight 150.0252
Chemical Name or Material 1H-Benzimidazole-2-thiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.