missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptobenzothiazole, 95%

Product Code. 10163460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
10g
1kg
250g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10163460 10 g Glass Bottle 10g
10494385 50 g Plastic bottle 50g
10716531 250 g Plastic bottle 250g
10522141 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10163460 Supplier Thermo Scientific Acros Supplier No. 125450100
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 149-30-4 | C7H5NS2 | 167.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-30-4
Molecular Formula C7H5NS2
Molecular Weight (g/mol) 167.24
MDL Number MFCD00005781
InChI Key YXIWHUQXZSMYRE-UHFFFAOYSA-N
Synonym 2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax
PubChem CID 697993
ChEBI CHEBI:34292
IUPAC Name 3H-1,3-benzothiazole-2-thione
SMILES C1=CC=C2C(=C1)NC(=S)S2

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 172.0°C to 182.0°C
Color Yellow
Flash Point 243°C
Loss on Drying 0.5% max. (60°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Quantity 10 g
Beilstein 27, II, 233
Merck Index 15, 5935
Solubility Information Solubility in water: 0.12g/L (25°C). Other solubilities: soluble in alkalies,soluble in alcohol,acetone,benzene and,chloroform
Formula Weight 167.24
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-Mercaptobenzothiazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.