missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptobenzothiazole, 97%

Product Code. 11418036
Click to view available options
Quantity:
10 g
100 g
500 g
2500 g
Unit Size:
100g
10g
2500g
500g
This item is not returnable. View return policy

Product Code. 11418036

Brand: Thermo Scientific Alfa Aesar A14086.0E

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 149-30-4 | C7H5NS2 | 167.244 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Mercaptobenzothiazole is used for detection of the equivalence point in volumetry (complexometry) and for concentration determinations in colorimetry. It is also used for the photometric determination of gold, cadmium, copper, lead and palladium. Further, it is used as a metalworking fluid, antifreeze, grease, anticorrosive agent, and adhesive. It acts as an antibacterial and antifungal. It is also employed in the rubber industry as a vulcanization accelerator. It inhibits dopamine beta-hydroxylase and biocorrosion in cooling systems, especially in the paper industry. In addition to this, it is used as a wood preservative and rubber additive.

Solubility
Soluble in ethanol, diethyl ether, benzene, carbon tetrachloride, alkali and alkaline solutions. Slightly soluble in acetone, naphtha and glacial acetic acid. Insoluble in water and gasoline.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149-30-4
Molecular Formula C7H5NS2
Molecular Weight (g/mol) 167.244
MDL Number MFCD00005781
InChI Key YXIWHUQXZSMYRE-UHFFFAOYSA-N
Synonym 2-mercaptobenzothiazole, 2-benzothiazolethiol, captax, benzothiazolethiol, 2 3h-benzothiazolethione, benzothiazole-2-thiol, 1,3-benzothiazole-2-thiol, benzo d thiazole-2-thiol, rokon, rotax
PubChem CID 697993
ChEBI CHEBI:34292
IUPAC Name 3H-1,3-benzothiazole-2-thione
SMILES C1=CC=C2C(=C1)NC(=S)S2

Specifications

Melting Point 178°C to 182°C
Density 1.42
Flash Point 243°C (469°F)
Quantity 2500 g
UN Number UN3077
Beilstein 119484
Sensitivity Air sensitive
Merck Index 14,5868
Solubility Information Soluble in ethanol,diethyl ether,benzene,carbon tetrachloride,alkali and alkaline solutions; Slightly soluble in acetone,naphtha and glacial acetic acid; Insoluble in water and gasoline.
Formula Weight 167.25
Percent Purity 97%
Chemical Name or Material 2-Mercaptobenzothiazole
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.