missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Mercaptoethanol, 99%, pure

Produktkode 10367100 Shop alle Thermo Scientific Chemicals produkter
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Förpackningsstorlek
1 L
10mL
2.5 L
250mL
Denne vare kan ikke returneres. Se returpolitik
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

Produktkode 10367100

missing translation for 'mfr': Thermo Scientific Acros 125472500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolitik
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

CAS: 60-24-2 | C2H6OS | 78.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 60-24-2
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00004890
InChI Key DGVVWUTYPXICAM-UHFFFAOYSA-N
Synonym 2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan
PubChem CID 1567
ChEBI CHEBI:41218
SMILES OCCS

Tekniske data

Melting Point -100.0°C
Color Colorless to Yellow
Density 1.1100g/mL
Boiling Point 157.0°C to 158.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula HSCH2CH2OH
Refractive Index 1.4990 to 1.5020
Quantity 250 mL
Beilstein 01,47
Fieser 01,643
Merck Index 15,5936
Specific Gravity 1.11
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol,ether,benzene,soluble in acetone,miscible with most common organic solvents
IUPAC Name 2-sulfanylethanol
Viscosity 3.43 mPa.s (20°C)
Formula Weight 78.13
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material 2-Mercaptoethanol
Visa mer Visa mindre
compliance-icons compliance-icons compliance-icons compliance-icons
Produktbeteckning
  • 2-Mercaptoethanol
Signalord
  • Danger
Riskkategorier
  • Acute toxicity Category 3
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
Faroangivelser
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H331-Toxic if inhaled.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H317-May cause an allergic skin reaction.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.