missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mercaptoethanol, 99%, pure
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
10mL
2.5 L
250mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -100.0°C |
| Color | Colorless to Yellow |
| Density | 1.1100g/mL |
| Boiling Point | 157.0°C to 158.0°C |
| Flash Point | 73°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | HSCH2CH2OH |
| Refractive Index | 1.4990 to 1.5020 |
| Show More |
Safety and Handling
Product Identifier
- 2-Mercaptoethanol
- Danger
- Acute toxicity Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1A
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- H301-Toxic if swallowed.
- H310-Fatal in contact with skin.
- H331-Toxic if inhaled.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H317-May cause an allergic skin reaction.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction