missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoethanol, Electrophoresis Grade, 98+%

Product Code. 15418219
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418219 250 g 250g
15400647 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418219 Supplier Thermo Scientific Alfa Aesar Supplier No. J66742.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60-24-2 | C2H6OS | 78.13 g/mol

β-Mercaptoethanol can act as an enzyme reactivator in systems necessitating reduction for activation, and has been commonly used to reduce disulfide bonds in order to separate protein subunits for use in electrophoresis. This product can also be used to form o-Phthalaldehyde-beta-mercaptoethanol (OPAME), a fluorogenic reagent shown to detect amino acids based on its reaction with primary amines. More recently OPAME has been used to distinguish between ssDNA/dsDNA and even single base mismatches in the hybridization of DNA. It is widely used for retarding oxidation of biological compounds in solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
β-Mercaptoethanol can act as an enzyme reactivator in systems necessitating reduction for activation, and has been commonly used to reduce disulfide bonds in order to separate protein subunits for use in electrophoresis. This product can also be used to form o-Phthalaldehyde-beta-mercaptoethanol (OPAME), a fluorogenic reagent shown to detect amino acids based on its reaction with primary amines. More recently OPAME has been used to distinguish between ssDNA/dsDNA and even single base mismatches in the hybridization of DNA. It is widely used for retarding oxidation of biological compounds in solution.

Solubility
Fully miscible in water. Soluble in alcohol, ether, benzene, and other organic solvents.

Notes
Handle under dry protective gas. Store in cool condition. Incompatible with air, moisture, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-24-2
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00004890
InChI Key DGVVWUTYPXICAM-UHFFFAOYSA-N
Synonym 2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan
PubChem CID 1567
ChEBI CHEBI:41218
SMILES OCCS

Specifications

Melting Point -100°C
Color Colorless
Density 1.114 g/mL
Boiling Point 156°C to 158°C
Flash Point 73°C (163°F)
Odor Unpleasant
Linear Formula HSCH2CH2OH
Quantity 250 g
UN Number UN2966
Beilstein 773648
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,5869
Solubility Information Fully miscible in water. Soluble in alcohol,ether,benzene,and other organic solvents.
IUPAC Name 2-sulfanylethanol
Formula Weight 78.13
Percent Purity ≥98%
Grade Electrophoresis
Physical Form Liquid
Chemical Name or Material 2-Mercaptoethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.