missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoethyl sulfide, 90%

Product Code. 12684937 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12684937 50 g Glass bottle 50g
12664937 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12684937 Supplier Thermo Scientific Acros Supplier No. 125490500
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 3570-55-6 | C4H10S3 | 154.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3570-55-6
Molecular Formula C4H10S3
Molecular Weight (g/mol) 154.30
MDL Number MFCD00004893
InChI Key KSJBMDCFYZKAFH-UHFFFAOYSA-N
Synonym 2,2'-thiodiethanethiol, bis 2-mercaptoethyl sulfide, 2-mercaptoethyl sulfide, ethanethiol, 2,2'-thiobis, bis mercaptoethyl sulfide, 3-thia-1,5-pentanedithiol, 2-mercaptoethylsulfide, 2,2'-dimercaptodiethyl sulfide, unii-oeu4azc07s, ethanethiol, 2,2'-thiodi
PubChem CID 77117
IUPAC Name 2-(2-sulfanylethylsulfanyl)ethanethiol
SMILES SCCSCCS

Specifications

Melting Point -11.0°C
Color Colorless
Density 1.1830g/mL
Boiling Point 135.0°C to 136.0°C (10.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 88 % min. (GC)
Packaging Glass bottle
Linear Formula (HSCH2CH2)2S
Refractive Index 1.594
Quantity 50 g
Specific Gravity 1.183
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,toluene,chloroform,,dioxane,carbon tetrachloride
Formula Weight 154.32
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 2-Mercaptoethyl sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.