missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoimidazole, 98+%

Product Code. 11324574
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324574 1 g 1g
11334574 5 g 5g
11344574 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324574 Supplier Thermo Scientific Alfa Aesar Supplier No. L01346.03
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 872-35-5 | C3H4N2S | 100.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-35-5
Molecular Formula C3H4N2S
Molecular Weight (g/mol) 100.14
MDL Number MFCD00005188
InChI Key OXFSTTJBVAAALW-UHFFFAOYSA-N
Synonym 2-mercaptoimidazole, 1h-imidazole-2-thiol, imidazole-2-thiol, 1h-imidazole-2 3h-thione, 4-imidazoline-2-thione, 1,3-vinylenethiourea, imidazole, 2-mercapto, 2h-imidazole-2-thione, 1,3-dihydro, usaf el-57, mercaptoimidazole
PubChem CID 1201386
SMILES SC1=NC=CN1

Specifications

Melting Point 222°C to 227°C
Quantity 1 g
Beilstein 105775
IUPAC Name 1H-imidazole-2-thiol
Formula Weight 100.14
Percent Purity ≥98%
Chemical Name or Material 2-Mercaptoimidazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.