missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoisobutyric Acid, 90%, TRC

Product Code. 30132733
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132733 10 g 10g
30134269 1 g 1g
30010857 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132733 Supplier Toronto Research Chemicals Supplier No. TRCM25340010G
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4695-31-2
Molecular Formula C4 H8 O2 S
Molecular Weight (g/mol) 120.17
IUPAC Name 2-methyl-2-sulfanylpropanoic acid
SMILES CC(C)(S)C(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C4H8O2S/c1-4(2,7)3(5)6/h7H,1-2H3,(H,5,6)
Formula Weight 120.0245
Chemical Name or Material 2-Mercaptoisobutyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.