missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyridine, 98%

Product Code. 11489273
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489273 5 g 5g
11499273 25 g 25g
11409283 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489273 Supplier Thermo Scientific Alfa Aesar Supplier No. A11741.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2637-34-5 | C5H5NS | 111.162 g/mol

Employed as a ligand in metal complexes. It can be used to coat porous media to purify plasmid DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a ligand in metal complexes. It can be used to coat porous media to purify plasmid DNA.

Solubility
Slightly soluble in chloroform, methanol and dimethyl sulfoxide.

Notes
Air Sensitive, keep the container tightly sealed. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2637-34-5
Molecular Formula C5H5NS
Molecular Weight (g/mol) 111.162
MDL Number MFCD00006285
InChI Key WHMDPDGBKYUEMW-UHFFFAOYSA-N
Synonym 2-mercaptopyridine, 2-pyridinethiol, pyridine-2-thiol, 2-thiopyridine, pyrid-2-thione, 2 1h-pyridinethione, pyridinethione, 2-thiopyridone, 2-pyridinethione, 2-mercapto pyridine
PubChem CID 2723698
ChEBI CHEBI:45223
IUPAC Name 1H-pyridine-2-thione
SMILES C1=CC(=S)NC=C1

Specifications

Melting Point 124°C to 130°C
Quantity 5 g
Beilstein 105758
Sensitivity Air sensitive
Solubility Information Slightly soluble in chloroform,methanol and dimethyl sulfoxide.
Formula Weight 111.17
Percent Purity 98%
Chemical Name or Material 2-Mercaptopyridine
compliance-icons
Product Identifier
  • 2-Mercaptopyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 2(1H)-Pyridinethione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.