missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyridine, 98%, Thermo Scientific Chemicals

Product Code. 10284930 Shop All Thermo Scientific Chemicals Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
2.5 g
25 g
100 g
Packaging:
Glass bottle
missing translation for 'unitSize':
100g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging missing translation for 'unitSize'
10284930 25 g Glass bottle 25g
10163220 2.5 g Glass bottle 2.5g
10783714 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10284930 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 125540250
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2637-34-5 | C5H5NS | 111.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2637-34-5
Molecular Formula C5H5NS
Molecular Weight (g/mol) 111.16
MDL Number MFCD00006285
InChI Key WHMDPDGBKYUEMW-UHFFFAOYSA-N
Synonym 2-mercaptopyridine, 2-pyridinethiol, pyridine-2-thiol, 2-thiopyridine, pyrid-2-thione, 2 1h-pyridinethione, pyridinethione, 2-thiopyridone, 2-pyridinethione, 2-mercapto pyridine
PubChem CID 2723698
ChEBI CHEBI:45223
IUPAC Name 1H-pyridine-2-thione
SMILES C1=CC(=S)NC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 126.0°C to 130.0°C
Color Brown to Yellow
Flash Point 125°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodimetry)
Packaging Glass bottle
Linear Formula (C5H4N)SH
Quantity 25 g
Beilstein 21,45
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: soluble in most common solvents
Formula Weight 111.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercaptopyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Mercaptopyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.