missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyridine-N-oxide, 99%

Product Code. 10609024 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10609024 25 g Glass bottle 25g
10467200 1 g Glass bottle 1g
10512401 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10609024 Supplier Thermo Scientific Acros Supplier No. 182670250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1121-31-9 | C5H5NOS | 127.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1121-31-9
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.16
MDL Number MFCD00006196
InChI Key YBBJKCMMCRQZMA-UHFFFAOYSA-N
Synonym pyrithione, 2-mercaptopyridine n-oxide, 2-mercaptopyridine-n-oxide, 1-hydroxy-2-pyridinethione, omadine, 2-pyridinethiol, 1-oxide, 1-hydroxyl-1h-pyridine-2-thione, n-hydroxypyridine-2-thione, 1-hydroxy-2 1h-pyridinethione, 2-pyridinethiol 1-oxide
PubChem CID 1570
ChEBI CHEBI:36578
IUPAC Name 1-hydroxypyridine-2-thione
SMILES C1=CC(=S)N(C=C1)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 70°C to 73°C
Color Beige to White or Gray-Green or Green-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Iodometry)
Packaging Glass bottle
Quantity 25 g
Beilstein 21, V,7, 150
Fieser 12,417; 14,268; 17,292
Merck Index 14, 7994
Formula Weight 127.16
Percent Purity 99%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 2-Mercaptopyridine-N-oxide
Show More Show Less
compliance-icons
Product Identifier
  • 2-Mercaptopyridine-N-oxide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.