missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyrimidine, 98%

Product Code. 10327620 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10327620 50 g Glass bottle 50g
10061670 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10327620 Supplier Thermo Scientific Acros Supplier No. 156740500
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1450-85-7 | C4H4N2S | 112.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1450-85-7
Molecular Formula C4H4N2S
Molecular Weight (g/mol) 112.15
MDL Number MFCD00006073
InChI Key HBCQSNAFLVXVAY-UHFFFAOYSA-N
Synonym 2-mercaptopyrimidine, pyrimidine-2-thiol, 2-pyrimidinethiol, 2-thiopyrimidine, pyrimidine-2 1h-thione, 2 1h-pyrimidinethione, pyrimidinethione, 2-pyrimidinethione, 2-pyrimidinethiol 9ci, 2-1h-pyrimidinthione
PubChem CID 1550489
ChEBI CHEBI:53576
IUPAC Name 1H-pyrimidine-2-thione
SMILES C1=CNC(=S)N=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 230°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 50 g
Formula Weight 112.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Mercaptopyrimidine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.