missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyrimidine, 98%

Product Code. 11475373
Change view
Click to view available options
Quantity:
10 g
100 g
250 g
Unit Size:
100g
10g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475373 250 g 250g
11465373 10 g 10g
11369589 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11475373 Supplier Thermo Scientific Alfa Aesar Supplier No. A13382.30
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 1450-85-7 | C4H4N2S | 112.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1450-85-7
Molecular Formula C4H4N2S
Molecular Weight (g/mol) 112.15
MDL Number MFCD00006073
InChI Key HBCQSNAFLVXVAY-UHFFFAOYSA-N
Synonym 2-mercaptopyrimidine, pyrimidine-2-thiol, 2-pyrimidinethiol, 2-thiopyrimidine, pyrimidine-2 1h-thione, 2 1h-pyrimidinethione, pyrimidinethione, 2-pyrimidinethione, 2-pyrimidinethiol 9ci, 2-1h-pyrimidinthione
PubChem CID 1550489
ChEBI CHEBI:53576
IUPAC Name 1H-pyrimidine-2-thione
SMILES C1=CNC(=S)N=C1

Specifications

Melting Point ∼230°C (decomposition)
Quantity 250 g
Beilstein 107105
Formula Weight 112.15
Percent Purity 98%
Chemical Name or Material 2-Mercaptopyrimidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.