missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptothiazoline, 98+%

Product Code. 11321767
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11321767 100 g 100g
11331767 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11321767 Supplier Thermo Scientific Alfa Aesar Supplier No. L04924.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 96-53-7 | C3H5NS2 | 119.2 g/mol

2-Mercaptothiazoline, is used in the chain-lengthening of alkyl halides by trans-iodopropenylenation. 2-Mercaptothiazoline is an antithyroid agent that strongly reduces thyroid hormone levels. 2-Mercaptothiazoline is also used as an intermediate in the preparation of biologically active compounds. It is also used as a tool for highly selective chiral synthesis of penam- and carbapenam-type β-lactam antibiotics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercaptothiazoline, is used in the chain-lengthening of alkyl halides by trans-iodopropenylenation. 2-Mercaptothiazoline is an antithyroid agent that strongly reduces thyroid hormone levels. 2-Mercaptothiazoline is also used as an intermediate in the preparation of biologically active compounds. It is also used as a tool for highly selective chiral synthesis of penam- and carbapenam-type β-lactam antibiotics.

Solubility
Soluble in chloroform, methanol. Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-53-7
Molecular Formula C3H5NS2
Molecular Weight (g/mol) 119.2
MDL Number MFCD00126013
InChI Key WGJCBBASTRWVJL-UHFFFAOYSA-N
Synonym 2-mercaptothiazoline, 2-thiazolidinethione, 2-thiazoline-2-thiol, thiazolidine-2-thione, metabasal, thyroidan, 2-mercapto-2-thiazoline, mercaptothiazoline, 2-thiothiazolidone, 2-thiazolidenethione
PubChem CID 2723699
IUPAC Name 1,3-thiazolidine-2-thione
SMILES C1CSC(=S)N1

Specifications

Melting Point 103°C to 107°C
Quantity 100 g
UN Number UN2811
Beilstein 106332
Solubility Information Soluble in chloroform,methanol. Slightly soluble in water.
Formula Weight 119.21
Percent Purity ≥98%
Chemical Name or Material 2-Mercaptothiazoline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.