missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-3-methylpyrazine, 99%

Product Code. 11367975
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11367975

Brand: Thermo Scientific Alfa Aesar L08067.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2847-30-5 | C6H8N2O | 124.143 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2847-30-5
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
MDL Number MFCD00006127
InChI Key VKJIAEQRKBQLLA-UHFFFAOYSA-N
Synonym pyrazine, 2-methoxy-3-methyl, 2-methyl-3-methoxypyrazine, 2-methoxy-3-methyl-pyrazine, 2-methoxy-3-methyl pyrazine, unii-04o7cn9q85, pyrazine, 2-methoxy-3 or 5-methyl, pyrazine, 2-methyl-3-methoxy, almond pyrazine, 2-methoxy-3 or5-methylpyrazine, chocolate pyrazine a
PubChem CID 17898
IUPAC Name 2-methoxy-3-methylpyrazine
SMILES CC1=NC=CN=C1OC

Specifications

Density 1.07
Boiling Point 159°C
Flash Point 55°C (131°F)
Refractive Index 1.5065
Quantity 5 g
UN Number UN1993
Beilstein 878809
Formula Weight 124.14
Percent Purity 99%
Chemical Name or Material 2-Methoxy-3-methylpyrazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.