missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-3-pyridinecarboxaldehyde, 96%, Thermo Scientific™

Product Code. 11966391 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11966391 5 g Glass bottle 5g
11956391 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11966391 Supplier Thermo Scientific Acros Supplier No. 439780050
 Restricted Item
Estimated Shipment: 14-12-2026
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Kemiske identifikatorer

CAS 71255-09-9
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
InChI Key PIFFMIDNNWOQLK-UHFFFAOYSA-N
Synonym 2-methoxynicotinaldehyde, 2-methoxy-3-pyridinecarboxaldehyde, 2-methoxy-pyridine-3-carbaldehyde, 2-methoxypyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2-methoxy, 2-methoxy-3-pyridine carboxaldehyde, 3-pyridinecarboxaldehyde,2-methoxy, 2-methoxy-3-pyridinecarbaldehyde, pubchem15124, 2-methoxy-3-formyl-pyridine
PubChem CID 12533391
IUPAC Name 2-methoxypyridine-3-carbaldehyde
SMILES COC1=C(C=CC=N1)C=O

Tekniske data

CAS Min % 95.0
CAS Max % 100.0
Melting Point 25°C
Color Colorless to Yellow
Boiling Point 200°C to 201°C
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 137.14
Percent Purity 96%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2-Methoxy-3-pyridinecarboxaldehyde
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • 2-Methoxy-3-pyridinecarboxaldehyde
Signalord
  • Warning
Farekategori
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.