missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-methylphenol-3,5,6-d3,OD, CDN

Product Code. 30314146
Change view
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.50g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314146 0.5 g 0.50g
30310022 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30314146 Supplier CDN Supplier No. CDND69630.5G
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Methoxy-4-methylphenol-3,5,6-d3,OD
CAS 20189-08-6
Molecular Formula C8 D4 H6 O2
Quantity 0.5 g
Synonym p-Cres-2,3,5-d3-ol-d, 6-methoxy- (8 CI), OD-3,5,6-Trideutero-4-methylguaiacol, 2-Methoxy-4-methylphenol-3,5,6-d3,OD, 2-Methoxy-4-methylphenol-O,3,5,6-D4, 6-Methoxy-4-methylphenol-O,2,3,5-D4, 4-Methylguaiacol-O,3,5,6-d4, 2-Methoxy-p-cresol-d4, 4-(Hydroxy-d)-3-methoxytoluene-2,5,6-d3
InChI Formula InChI=1 S/C8H10O2/c1-6-3-4-7(9)8(5-6)10-2/h3-5,9 H,1-2H3/i3D,4 D,5 D/hD
SMILES [2 H]Oc1c([2 H])c([2 H])c(C)c([2 H])c1OC
IUPAC Name 1,2,5-trideuterio-3-deuteriooxy-4-methoxy-6-methylbenzene
Molecular Weight (g/mol) 142.1885
Formula Weight 142.0932 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.