missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-6-(tri-n-butylstannyl)pyrazine, 95%

Product Code. 15448117
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448117 250 mg 250mg
15438117 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448117 Supplier Thermo Scientific Alfa Aesar Supplier No. H51528.MD
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1105511-66-7 | C17H32N2OSn | 399.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1105511-66-7
Molecular Formula C17H32N2OSn
Molecular Weight (g/mol) 399.166
MDL Number MFCD08275735
InChI Key WWRXJELRWGUPKZ-UHFFFAOYSA-N
Synonym 2-methoxy-6-tributylstannyl pyrazine, 2-tri-n-butylstannyl-6-methoxypyrazine, 2-methoxy-6-tributylstannanyl pyrazine, 2-methoxy-6-tri-n-butylstannyl pyrazine, 2-tributylstannyl-6-methoxypyrazine
PubChem CID 16427103
IUPAC Name tributyl-(6-methoxypyrazin-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=NC(=CN=C1)OC

Specifications

Quantity 250 mg
UN Number UN2788
Formula Weight 399.15
Percent Purity 95%
Chemical Name or Material 2-Methoxy-6-(tri-n-butylstannyl)pyrazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.