missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxy-d3-pyrazine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | 2-Methoxypyrazine-D3 (methoxy-D3) |
| CAS | 32046-21-2 |
| Molecular Formula | C5 D3 H3 N2 O |
| Quantity | 0.1 g |
| Synonym | Pyrazine, methoxy-d3- (8 CI), 2-Methoxy-d3-pyrazine, 2-Methoxypyrazine-D3 (methoxy-D3), Methoxy-d3-pyrazine |
| InChI Formula | InChI=1 S/C5H6N2O/c1-8-5-4-6-2-3-7-5/h2-4 H,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])Oc1cnccn1 |
| IUPAC Name | 2-(trideuteriomethoxy)pyrazine |
| Molecular Weight (g/mol) | 113.1324 |
| Formula Weight | 113.0668 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction