missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzhydrazide, 98+%

Product Code. 11460184
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460184 50 g 50g
11450184 10 g 10g
11470184 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460184 Supplier Thermo Scientific Alfa Aesar Supplier No. A13619.18
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 7466-54-8 | C8H10N2O2 | 166.18 g/mol

2-Methoxybenzhydrazide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxybenzhydrazide is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7466-54-8
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00014755
InChI Key QMGXWNSSMGAHCA-UHFFFAOYSA-N
Synonym 2-methoxybenzhydrazide, 2-methoxybenzoic acid hydrazide, o-methoxybenzohydrazide, o-methoxybenzoylhydrazine, 2-methoxybenzoylhydrazide, 2-methoxybenzoylhydrazine, o-anisohydrazide, hydrazine, 2-methoxybenzoyl, o-methoxybenzoylhydrazide, o-methoxybenzoic acid hydrazide
PubChem CID 24051
IUPAC Name 2-methoxybenzohydrazide
SMILES COC1=CC=CC=C1C(=O)NN

Specifications

Melting Point 78°C to 82°C
Quantity 50 g
Beilstein 2208367
Solubility Information Slightly soluble in water.
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 2-Methoxybenzhydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.