missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxybenzoyl Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Methoxybenzoyl Chloride |
| CAS | 21615-34-9 |
| Molecular Formula | C8H7ClO2 |
| Quantity | 250 g |
| Synonym | o-Anisoyl Chloride, 2-(Methyloxy)benzoyl Chloride, 2-Anisoyl Chloride, 2-Methoxybenzoic Acid Chloride, 2-Methoxybenzoyl Chloride, o-Methoxybenzoyl Chloride, ortho-Anisoyl Chloride |
| InChI Formula | InChI=1S/C8H7ClO2/c1-11-7-5-3-2-4-6(7)8(9)10/h2-5H,1H3 |
| SMILES | COc1ccccc1C(=O)Cl |
| Molecular Weight (g/mol) | 170.59 |
| Formula Weight | 170.0135 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?