missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzylmagnesium chloride, 0.25M in 2-MeTHF

Product Code. 15400866
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15400866 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15400866 Supplier Thermo Scientific Alfa Aesar Supplier No. H54177.AE
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 480438-46-8 | C8H9ClMgO | 180.91 g/mol

2-Methoxybenzylmagnesium chloride is a Grignard reagent used in organic synthesis. It is involved in the synthesis of (-)-Chamobtusin A. It is also used in the preparation of molybdenum and tungsten Alkylidene complexes having a tert-butylimido ligand.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxybenzylmagnesium chloride is a Grignard reagent used in organic synthesis. It is involved in the synthesis of (-)-Chamobtusin A. It is also used in the preparation of molybdenum and tungsten Alkylidene complexes having a ter
TRUSTED_SUSTAINABILITY

Specifications

CAS 480438-46-8
Molecular Formula C8H9ClMgO
MDL Number MFCD01319897
Quantity 100 mL
UN Number UN2924
Synonym 2-methoxybenzylmagnesium chloride, 2-methoxybenzylmagnesium chloride solution, 2-methoxybenzyl magnesium chloride, 2-methoxybenzyl-magnesium chloride, jjnlsbbaaqtdsm-uhfffaoysa-m, magnesium,1-methanidyl-2-methoxybenzene,chloride, 2-methoxybenzylmagnesium chloride, 0.25m in 2-methf, 2-methoxybenzylmagnesium chloride 0.25 m in tetrahydrofuran, 2-methoxybenzylmagnesium chloride solution, 0.25 m in thf
InChI Key JJNLSBBAAQTDSM-UHFFFAOYSA-M
SMILES COC1=CC=CC=C1C[Mg]Cl
IUPAC Name magnesium;1-methanidyl-2-methoxybenzene;chloride
Molecular Weight (g/mol) 180.91
PubChem CID 13004954
Formula Weight 180.92
Concentration or Composition (by Analyte or Components) 0.25M in 2-MeTHF
Chemical Name or Material 2-Methoxybenzylmagnesium chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.