missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 2-Methoxyestradiol

Product Code. 16424938
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16424938 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16424938 Supplier MedChemExpress Supplier No. HY1203310mg
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 2-Methoxyestradiol (2-ME2), an orally active endogenous metabolite of 17β-estradiol (E2), is an apoptosis inducer and an angiogenesis inhibitor with potent antineoplastic activity. 2-Methoxyestradiol also destablize microtubules. 2-Methoxyestradio, also a potent superoxide dismutase (SOD) inhibitor and a ROS-generating agent, induces autophagy in the transformed cell line HEK293 and the cancer cell lines U87 and HeLa.

Specifications

Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 302.41 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-programmed cell death
CAS 362-07-2
Product Type Natural Products
Purity 96.96%
Solubility DMSO : ≥ 100 mg/mL (330.68 mM) ∣H2O : < 0.1 mg/mL (insoluble)
Synonym 2-ME2∣ NSC-659853
Target Microtubule/Tubulin∣Reactive Oxygen Species∣Apoptosis∣Autophagy∣Endogenous Metabolite
Quantity 10 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.