missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethanol, 99+%, for spectroscopy

Product Code. 10616762 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
Product Code. Quantity Packaging unitSize
10616762 2.5 L Glass bottle 2.5 L
10052100 1 L Glass bottle 1 L
2 options
This item is not returnable. View return policy

Product Code. 10616762

Brand: Thermo Scientific Acros 168580025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-86-4 | C3H8O2 | 76.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-86-4
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.09
MDL Number MFCD00002867
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
IUPAC Name 2-methoxyethanol
SMILES COCCO

Specifications

Acidity 0.0008 meq/g max.
Absorbance (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.05 max. at 270nm, 0.05 max. at 280nm, 0.1 max. at 260nm, 0.15 max. at 250nm, 0.2 max. at 240nm, 0.4 max. at 230nm
CAS Min % 99.0
CAS Max % 100.0
Melting Point -85.0°C
Density 0.9600g/mL
Boiling Point 124.0°C
Flash Point 38°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3OCH2CH2OH
Refractive Index 1.402
Quantity 2.5 L
Beilstein 01, 467
Residue after Evaporation 0.001% max.
Merck Index 15, 6063
Specific Gravity 0.96
Solubility Information Solubility in water: soluble.
Viscosity 1.7 mPa.s (20°C)
Formula Weight 76.09
Percent Purity 99+%
Grade Spectroscopy
Physical Form Liquid
Chemical Name or Material 2-Methoxyethanol, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.