Learn More
2-Methoxyethanol, 99%
Description
Solvent for ninhydrin color reagent2-Methoxy ethanol is used as a solvent for various purposes like varnishes, dyes, cleaning agents, epoxy coating, paints, anti-icing fuel additive in aviation and resins. It is involved in the preparation of Vaska’s complex viz. trans-chlorocarbonylbis(triphenylphosphine)iridium(I) and ruthenium hydridechlorocarbonyl tristriphenylphosphine. Further, it is also used in the synthesis of 2-aminothiazoles and Rho-associated kinase inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Solvent for ninhydrin color reagent2-Methoxy ethanol is used as a solvent for various purposes like varnishes, dyes, cleaning agents, epoxy coating, paints, anti-icing fuel additive in aviation and resins. It is involved in the preparation of Vaska’s complex viz. trans-chlorocarbonylbis(triphenylphosphine)iridium(I) and ruthenium hydridechlorocarbonyl tristriphenylphosphine. Further, it is also used in the synthesis of 2-aminothiazoles and Rho-associated kinase inhibitors.
Solubility
Miscible with water, alcohol, ether, acetone, hydrocarbons, ketones, glycols and dimethylformamide.
Notes
Incompatible with strong bases, strong oxidizing agents, acids, bases, copper alloys and copper.
Specifications
Specifications
| Melting Point | -85°C |
| Density | 0.965 |
| pH | 4 to 7 |
| Boiling Point | 124°C to 125°C |
| Flash Point | 46°C (115°F) |
| Odor | Characteristic |
| Linear Formula | CH3OCH2CH2OH |
| Refractive Index | 1.4025 |
| Quantity | 500 mL |
| UN Number | UN1188 |
| Show More |
Safety and Handling
- 2-Methoxyethanol
- Danger
- Acute toxicity Category 4
- Flammable liquid Category 3
- Reproductive toxicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity after single exposure Category 1
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H360FD-May damage fertility. May damage the unborn child.
- H370-Causes damage to organs.
- H373-May cause damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : 2-Methoxyethanol
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.