Learn More
2-Methoxyethanol, ACS, 99.3+%
Description
2-Methoxy ethanol is used as solvent for various purposes like varnishes, dyes, cleaning agents, epoxy coating, paints, anti-icing fuel additive in aviation and resins. It is involved in the preparation of Vaska’s complex viz. trans-chlorocarbonylbis(triphenylphosphine)iridium(I) and ruthenium hydridechlorocarbonyl tristriphenylphosphine. Further, it is also used in the synthesis of 2-aminothiazoles and Rho-associated kinase inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | °C to 85°C |
| Density | 0.965 |
| Boiling Point | 124°C to 125°C |
| Flash Point | 46°C (115°F) |
| Assay Percent Range | 99.3+% |
| Refractive Index | 1.4025 |
| Quantity | 250 mL |
| Beilstein | 1731074 |
| Solubility Information | Miscible with water,alcohol,glycerol,ether,acetone,DMFMiscible with water,alcohol,ether,acetone,hydrocarbons,ketones,glycols and dimethylformamide. |
| Formula Weight | 76.1 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.