missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethanol, ACS, 99.3+%

Product Code. 11328277
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
Product Code. Quantity unitSize
11328277 1 L 1 L
11318277 250 mL 250mL
2 options
This item is not returnable. View return policy

Product Code. 11328277

Brand: Thermo Scientific Alfa Aesar 031733.K2

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-86-4 | C3H8O2 | 76.095 g/mol

2-Methoxy ethanol is used as solvent for various purposes like varnishes, dyes, cleaning agents, epoxy coating, paints, anti-icing fuel additive in aviation and resins. It is involved in the preparation of Vaska’s complex viz. trans-chlorocarbonylbis(triphenylphosphine)iridium(I) and ruthenium hydridechlorocarbonyl tristriphenylphosphine. Further, it is also used in the synthesis of 2-aminothiazoles and Rho-associated kinase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-86-4
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
MDL Number MFCD00002867
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
IUPAC Name 2-methoxyethanol
SMILES COCCO

Specifications

Melting Point °C to 85°C
Density 0.965
Boiling Point 124°C to 125°C
Flash Point 46°C (115°F)
Assay Percent Range 99.3+%
Refractive Index 1.4025
Quantity 1 L
Beilstein 1731074
Solubility Information Miscible with water,alcohol,glycerol,ether,acetone,DMFMiscible with water,alcohol,ether,acetone,hydrocarbons,ketones,glycols and dimethylformamide.
Formula Weight 76.1
Grade ACS
Physical Form Liquid
Chemical Name or Material 2-Methoxyethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.