missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethanol, ACS reagent

Product Code. 11914751 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11914751 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11914751 Supplier Thermo Scientific Acros Supplier No. 447480010
  • ONLINE EXCLUSIVE PRICE
    74.40 €
    68.40 € / 1 L
    Save 6.00 €
    8% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 109-86-4 | C3H8O2 | 76.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 109-86-4
Melting Point -85.0°C
Density 0.9600g/mL
Boiling Point 124.0°C
Flash Point 38°C
Molecular Formula C3H8O2
Linear Formula CH3OCH2CH2OH
Refractive Index 1.402
Quantity 1 L
Merck Index 15,6063
Specific Gravity 0.96
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
Solubility Information Solubility in water: soluble
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILES COCCO
IUPAC Name 2-methoxyethanol
Molecular Weight (g/mol) 76.09
PubChem CID 8019
ChEBI CHEBI:46790
Viscosity 1.7 mPa.s (20°C)
Formula Weight 76.09
Grade ACS Reagent
Titratable Acid 0.002 meq/g max
Chemical Name or Material 2-Methoxyethanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methoxyethanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H370-Causes damage to organs.
  • H360FD-May damage fertility. May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.